Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB100479X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100479X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100479X Supplier Novus Biologicals Supplier No. NB100479AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Canine, Fish, Goat, Hamster, Human, Mouse, Pig, Primate, Rabbit, Rat, Zebrafish samples. It is validated for ChIP Assay, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Immunoprecipitation, Western Blot.

Specifications

Antigen HIF-1 alpha
Applications ChIP Assay, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Gene Symbols HIF1A
Host Species Rabbit
Immunogen This HIF-1 alpha Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221].
Molecular Weight of Antigen 93 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091
Test Specificity Human, mouse, rat, goat, primate and rabbit. Hamster reactivity reported in scientific literature (PMID: 24997364 and 24997360).
Target Species Canine, Fish, Goat, Hamster, Human, Mouse, Pig, Primate, Rabbit, Rat, Zebrafish
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.