Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB355142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355142 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355142 Supplier Novus Biologicals Supplier No. NB100134APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Bovine, Canine, Guinea Pig, Human, Mouse, Primate, Rat, Xenopus species, Zebrafish samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), Dual RNAScope ISH-IHC, ELISA, Gel Super Shift Assays, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen HIF-1 alpha
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dual RNAScope ISH-IHC, ELISA, Gel Supershift, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing, Dual RNAScope ISH-IHC
Formulation PBS
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8
Host Species Rabbit
Immunogen This HIF-1 alpha Antibody was developed against a fusion protein made to an internal sequence of human HIF-1 alpha (containing amino acids 432 - 528) [Uniprot# Q16665].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Cytokine Research, HIF Target Genes, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091
Target Species Bovine, Canine, Guinea Pig, Human, Mouse, Primate, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.