Learn More
Description
Specifications
Specifications
| Antigen | HIF-1 alpha |
| Applications | ChIP Assay, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, KnockDown, Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2ug/ml, Simple Western, Flow Cytometry 1:10 - 1:1000. Use reported in scientific literature (PMID 21917971), Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Immunohistochemistry-Frozen 5 - 10 ug/mL, Immunoblotting reported in scientific literature (PMID 28506759), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 27148974), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q61221 |
| Gene Alias | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp |
| Gene Symbols | HIF1A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
