Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100654 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100654 0.1 mL
NB100654SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100654 Supplier Novus Biologicals Supplier No. NB100654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Bovine, Human, Mouse, Pig, Rat samples. It is validated for Gel Super Shift Assays, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot.

Specifications

Antigen HIF-1 alpha
Applications Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:100 - 1:300. Use reported in scientific literature, Immunohistochemistry-Frozen 1:100 - 1:300. Use reported in scientific literature, Gel Super Shift Assays, KnockDown Validated reported in scientific literature (PMID 32054096)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q16665
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Gene Symbols HIF1A
Host Species Rabbit
Immunogen This HIF-1 alpha Antibody was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665].
Molecular Weight of Antigen 93 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091
Test Specificity Human and porcine. Does not react with rat. Immunogen sequence has 93% homology to bovine and 90% homology to mouse.Mouse reactivity reported in scientific literature (PMID: 23959856) Rat reactivity reported in scientific literature (PMID: 24122166).
Target Species Bovine, Mouse, Human, Mouse, Pig, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.