Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB100134UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100134UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100134UV Supplier Novus Biologicals Supplier No. NB100134UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Bovine, Canine, Guinea Pig, Human, Mouse, Primate, Rat, Xenopus species, Zebrafish samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), Dual RNAScope ISH-IHC, ELISA, Gel Super Shift Assays, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen HIF-1 alpha
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dual RNAScope ISH-IHC, ELISA, Gel Supershift, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Gene Symbols HIF1A
Host Species Rabbit
Immunogen This HIF-1 alpha Antibody was developed against a fusion protein made to an internal sequence of human HIF-1 alpha (containing amino acids 432 - 528) [Uniprot# Q16665].
Molecular Weight of Antigen 93 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091.0
Target Species Bovine, Canine, Guinea Pig, Human, Mouse, Primate, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.