Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody (ESEE122), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 81 publication
$244.50 - $560.50
Specifications
Antigen | HIF-1 alpha |
---|---|
Clone | ESEE122 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Simple Western 1:2000, Flow Cytometry reported in scientific literature (Gestier S. et al), Immunohistochemistry 1:100-1:5000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 26757928 Fig1G), Immunohistochemistry-Paraffin 1:100-1:5000, Immunohistochemistry-Frozen 1:100-1:5000, Immunoblotting |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100131SS
![]() |
Novus Biologicals
NB100131SS |
0.025 mL |
Each for $244.50
|
|
|||||
NB100131
![]() |
Novus Biologicals
NB100131 |
0.1 mL |
Each for $560.50
|
|
|||||
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
HIF-1 alpha | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
Q16665 | |
3091 | |
This HIF-1 alpha Antibody (ESEE122) was developed against Human HIF-1 alpha, corresponding to amino acids 329 - 530 [Uniprot# Q16665]. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
93 kDa |
ESEE122 | |
Western Blot 1:500-1:1000, Simple Western 1:2000, Flow Cytometry reported in scientific literature (Gestier S. et al), Immunohistochemistry 1:100-1:5000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 26757928 Fig1G), Immunohistochemistry-Paraffin 1:100-1:5000, Immunohistochemistry-Frozen 1:100-1:5000, Immunoblotting | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Canine | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
HIF1A | |
IgG1 | |
Protein G purified | |
Stains HIF-1 alpha in free-floating canine brain sections, according to customer reports. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title