Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody (H1alpha67), Novus Biologicals™
SDP

Catalog No. NB100123SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100123SS 0.025 mL
NB100123 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100123SS Supplier Novus Biologicals Supplier No. NB100123SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Avian, Bovine, Canine, Ferret, Human, Mouse, Ovine, Pig, Primate, Rabbit, Rat, Sheep samples. It is validated for Chemotaxis, ChIP Assay, ELISA, Flow Cytometry, Gel Super Shift Assays, Gel Supershift, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot.

Specifications

Antigen HIF-1 alpha
Applications Chemotaxis, ChIP Assay, ELISA, Flow Cytometry, Gel Supershift, Gel Supershift, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot
Classification Monoclonal
Clone H1alpha67
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000. Use reported in scientific literature, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10, Immunohistochemistry-Paraffin 1:100 - 1:300, Immunoblotting reported in multiple pieces of scientific literature, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 27991597), KnockDown Validated
Gene Accession No. Q16665
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1
Gene Symbols HIF1A
Host Species Mouse
Immunogen This HIF-1 alpha Antibody (H1alpha67) was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665].
Molecular Weight of Antigen 93 kDa
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091
Test Specificity Recognizes human, mouse, rat, bovine, monkey, porcine, sheep, canine, bird and ferret HIF-1 alpha.
Target Species Avian, Mouse, Bovine, Canine, Ferret, Bovine, Human, Ferret, Mouse, Sheep, Ovine, Pig, Primate, Rabbit, Rat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.