Learn More
Description
Specifications
Specifications
| Antigen | HIF-2 alpha/EPAS1 |
| Applications | Blocking Assay, Chemotaxis, ChIP Assay, Co-immunoprecipitation (Co-IP), Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Supershift, Gel Supershift, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, In vivo Assay, KnockDown, SDS-Page, Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 ug/mL, Simple Western 1:50, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 5 ug/ 1 mg lysate, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28115701), In vitro assay reported in scientific literature (PMID 24998849), Gel Super Shift Assays reported in scientific literature (PMID 15184875), SDS-Page, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 26861754), KnockDown Validated reported in scientific literature (PMID 31061092), Dual RNAscope ISH-IHC |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | Q99814 |
| Gene Alias | anti hif 2 alpha, anti-HIF-2 alpha, Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, |
| Gene Symbols | EPAS1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
