Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-2 alpha/EPAS1 Antibody (ep190b), Novus Biologicals™
SDP

Catalog No. NB100132SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100132SS 0.025 mL
NB100132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100132SS Supplier Novus Biologicals Supplier No. NB100132SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 177 publications

HIF-2 alpha/EPAS1 Monoclonal specifically detects HIF-2 alpha/EPAS1 in Bovine, Hamster, Human, Mouse, Rat samples. It is validated for Chemotaxis, ChIP Assay, ELISA, EMSA, Flow Cytometry, Gel Super Shift Assays, Gel Supershift, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, KnockDown, Simple Western, Western Blot.

Specifications

Antigen HIF-2 alpha/EPAS1
Applications Chemotaxis, ChIP Assay, ELISA, EMSA, Flow Cytometry, Gel Supershift, Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, KnockDown, Simple Western, Western Blot
Classification Monoclonal
Clone ep190b
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Simple Western 1:100, Flow Cytometry 1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:150 - 1:300, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:150 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24973414), Gel Super Shift Assays reported in scientific literature (PMID 17404621), In vivo assay reported in scientific literature (PMID 23857308), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated reported in scientific literature (PMID 32054096)
Gene Accession No. Q99814
Gene Alias Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, ep190b, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-
Gene Symbols EPAS1
Host Species Mouse
Immunogen The immunogen recognized by this HIF-2 alpha/EPAS1 Antibody (ep190b) maps to a region between amino acids 535-631. [UniProt# Q99814]
Molecular Weight of Antigen 96.5 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2034
Test Specificity This HIF-2 alpha/EPAS1 Antibody (ep190b) is specific for HIF-2 alpha/EPAS1, and does not cross-react with HIF-1 alpha.
Target Species Bovine, Mouse, Hamster, Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.