Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-2 alpha/EPAS1 Antibody, PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N10122PECY5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10122PECY5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10122PECY5 Supplier Novus Biologicals Supplier No. NB100122PECY55
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Fish, Hamster, Human, Mouse, Primate, Rabbit, Rat, Reptile, Sheep samples. It is validated for ChIP Assay, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Super Shift Assays, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, SDS-Page, Western Blot.

Specifications

Antigen HIF-2 alpha/EPAS1
Applications ChIP Assay, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Supershift, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, SDS-Page, Western Blot
Classification Polyclonal
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry, Knockout Validated, Knockdown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha,
Gene Symbols EPAS1
Host Species Rabbit
Immunogen This HIF-2 alpha/EPAS1 Antibody was developed against a peptide derived from the C-terminus of mouse/human HIF-2 alpha protein.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, Embryonic Stem Cell Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2034.0
Test Specificity This HIF-2 alpha/EPAS1 Antibody is specific for HIF-2 alpha/EPAS, and does not cross-react with HIF-1 alpha.
Target Species Fish, Hamster, Human, Mouse, Primate, Rabbit, Rat, Reptile, Sheep
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.