Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-3 alpha Antibody (OTI2D2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288353 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288353 Supplier Novus Biologicals Supplier No. NBP272136APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIF-3 alpha Monoclonal antibody specifically detects HIF-3 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen HIF-3 alpha
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D2
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Basic-helix-loop-helix-PAS protein MOP7, BHLHE17, bHLHe17HIF-3A2, Class E basic helix-loop-helix protein 17, HIF-3A4, HIF-3-alpha, HIF3-alpha, hypoxia inducible factor 3, alpha subunit, hypoxia-inducible factor 3-alpha, hypoxia-inducible factor-3 alpha 4, Inhibitory PAS domain protein, IPASHIF3-alpha-1, Member of PAS protein 7, MOP7PAS domain-containing protein 7, PASD7HIF-3A
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 355-669 of human HIF3A (NP_690008) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 64344
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.