Learn More
HIF2-alpha Recombinant Rabbit Monoclonal Antibody (BL-95-1A2), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $378.84
Specifications
Antigen | HIF2-alpha |
---|---|
Clone | BL-95-1A2 |
Applications | ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1260
![]() |
Bethyl Laboratories, Inc
A700003T |
20 μL |
Each for $157.08
|
|
|||||
50-157-1259
![]() |
Bethyl Laboratories, Inc
A700003 |
100 μL |
Each for $378.84
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.Specifications
HIF2-alpha | |
ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q99814 | |
2034 | |
Between 400 and 450 | |
Primary | |
4° C | |
EPAS1 |
BL-95-1A2 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
Hypoxia inducible Factor 2 alpha, MOP2, Endothelial pas domain protein 1, Member of pas superfamily 2 | |
EPAS1 | |
IgG | |
Ion-exchange chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.