Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIGD1A Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB164028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164028 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164028 Supplier Novus Biologicals Supplier No. NBP281721AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIGD1A Polyclonal antibody specifically detects HIGD1A in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen HIGD1A
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias 7420700H20Rik, DKFZP564K247, HIG1 domain family member 1A, HIG1 domain family, member 1A, HIG1 hypoxia inducible domain family, member 1A, HIG1DKFZp564K247, hypoxia inducible gene 1,2210020B17Rik, Hypoxia-inducible gene 1 protein
Host Species Rabbit
Immunogen HIGD1A antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human HIGD1A. The immunogen is located within the first 50 amino acids of HIGD1A. Amino Acid Squence: SLPSYEEDQGSKLIRKAKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 25994
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.