Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIGD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560331
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA560331 100 μL
1 options

Catalog No. PIPA560331

Supplier: Invitrogen™ PA560331

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STDTGVSLPS YEEDQGSKLI RKAKEAPFV Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 69%.

Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD1A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HIGD1A
Gene Accession No. Q9Y241
Gene Alias 2210020B17Rik; 7420700H20Rik; AI303276; AW049839; DKFZP564K247; Hig1; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family member 1A; HIG1 hypoxia inducible domain family, member 1A; Higd1a; HIMP1; HSPC010; hypoxia induced gene 1; hypoxia inducible gene 1; hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a
Gene Symbols HIGD1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HIGD1A. Recombinant protein control fragment (Product #RP-100253).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.