Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIGD2A Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB164065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164065 Supplier Novus Biologicals Supplier No. NBP281722JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIGD2A Polyclonal antibody specifically detects HIGD2A in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen HIGD2A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias HIG1 domain family member 2A, HIG1 domain family, member 2A, HIG1 hypoxia inducible domain family, member 2A, MGC2198
Host Species Rabbit
Immunogen HIGD2A antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human HIGD2A. The immunogen is located within the last 50 amino acids of HIGD2A. Amino Acid Squence: LANTEPTKGLPDHPSR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 192286
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.