Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ High Endothelial Venule Marker Monoclonal Antibody (MECA-79), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 50246086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-086 100 μg
50-246-085 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-246-086 Supplier Invitrogen™ Supplier No. 53603682
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This MECA-79 monoclonal antibody recognizes a sulfated carbohydrate epitope present on human and mouse high endothelial venules (HEVs) called peripheral lymph node addressin (PNAd). This glycoprotein is expressed on the luminal surface and in the cytoplasm of HEVs in lymphoid tissues, including peripheral and mesenteric lymph nodes and, to a lesser extent, Peyer's patches. The MECA-79 antigen has also been detected on follicular B cells in the spleen and appendix in humans, as well as at sites of chronic inflammation. Post-capillary venules or large vessels in the spleen, thymus, or non-lymphoid tissues do not express the MECA-79 antigen and thus cannot be detected with this antibody. This addressin is involved in lymphocyte infiltration and adhesion into secondary lymphoid tissues. The MECA-79 antibody has been reported to inhibit CD62L (L-selectin)-dependent lymphocyte emigration to HEVs and adhesion in lymph nodes and tonsils. Applications Reported: This MECA-79 antibody has been reported for use in flow cytometric analysis and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections (IHC-P). Applications Tested: This MECA-79 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded (FFPE) human tonsil with low pH antigen retrieval at less than or equal to 20 μg/mL. It is recommended that the anitbody be titrated for optimal performance in the assay of interest.

HEV peripheral lymph node addressin (PNAd) is a glycoprotein is expressed on the luminal surface and in the cytoplasm of HEVs in lymphoid tissues, including peripheral and mesenteric lymph nodes and, to a lesser extent, Peyer's patches. The MECA-79 antigen has also been detected on follicular B cells in the spleen and appendix in humans, as well as at sites of chronic inflammation. Post-capillary venules or large vessels in the spleen, thymus, or non-lymphoid tissues do not express the MECA-79 antigen and thus cannot be detected with this antibody. This addressin is involved in lymphocyte infiltration and adhesion into secondary lymphoid tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen High Endothelial Venule Marker
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone MECA-79
Concentration 0.5 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias HEV; MECA-79; PNAd
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.