Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High Five™ SMS Disposable Lab Coats
Shop All Ansell Products
Click to view available options
Color:
Blue
Purple
White
Size:
2X-Large
3X-Large
4X-Large
6X-Large
Large
Large/X-Large
Medium
Small
X-Large
Description
Breathable for all day comfort
- Protect against dry particulates and water-based liquids
- Knit collar and cuffs
- Five-snap front closure
- Available in white or blue

Specifications
Specifications
Product Type | General Purpose Lab Coat |
Size | 2X-Large |
Gender | Unisex |
Material | SMS |
Color | Purple |
Closure Type | Snaps |
No. of Pockets | 3 |
For Use With (Application) | Protects from dust, dirt, grime, and water based liquids |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction