Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Spunbond Isolation Gowns

Catalog No. 19156241 Shop All Ansell Products
Click to view available options
Color:
Blue
Pink
White
Yellow
Size:
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Durable protection from common dirt, dust and grime, minor splashes and particulates

Spunbond polypropylene material protects arms, chest, shoulders, waist and upper legs from common workplace particulates.

  • Lightweight, breathable fabric keeps workers cool and comfortable
  • Elastic wrists with easy neck and waist ties for wrap-around styling

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protection from common dirt, dust and grime, minor splashes and particulates
Product Type General Purpose Gown
Gender Unisex
Size 2X-Large
No. of Pockets None
Material Spunbond Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Model AG185
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.