Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ PPSB Lab Coats

Catalog No. p-4598895 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-156-162 Medium
19-156-164 X-Large
19-156-163 Large
19-156-165 2X-Large
19-156-168 5X-Large
19-156-166 3X-Large
19-156-167 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-156-162 Supplier High Five™ Supplier No. EW10203M
This item is not returnable. View return policy

Hook-and-loop closure for an adjustable fit

  • Breathable and comfortable spunbound polypropylene
  • Protects from dust and grime
  • Closure stays securely shut
  • White

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Hook and Loop
For Use With (Application) Protection from dust, dirt, and grime
Construction Breathable and Comfortable Spunbond Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.