Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva High Purity dNTPs

Used for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling. Cytiva High Purity dNTPs are tested functionally in long PCR* to be PCR-quantified for high-quality sequencing.

$763.40 - $947.10

Specifications

Format Solid
Content And Storage -20°C
Purity ≥98% triphosphate
Quantity 250 mg
For Use With (Application) Amplification, Dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
45-001-333
View Documents
Cytiva
27185004
dATP
Each for $763.40
Only null left
Add to Cart
 
45-001-335
View Documents
Cytiva
27187004
dGTP
Each for $947.10
Only null left
Add to Cart
 
45-001-334
View Documents
Cytiva
27186004
dCTP
Each for $947.10
Only null left
Add to Cart
 
45-002-407
View Documents
Cytiva
27188004
dTTP
Each for $789.80
Only null left
Add to Cart
 
Description

Description

  • High purity: >98% triphosphate, as confirmed by chromatographic analysis and 31P nuclear magnetic resonance (NMR) spectroscopy
  • Supplied as solids and also available in solutions
Specifications

Specifications

Solid
≥98% triphosphate
Amplification, Dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling
-20°C
250 mg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.