Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Petrifilm™ High-Sensitivity Coliform Count Plate, Sold by AquaPhoenix Scientific
Accommodates 5 mL samples and detection of low levels of confirmed coliforms in 24 hours
Specifications
Detects | Coliform |
---|---|
Product Type | Count Plate |
Test Type | Environmental Test |
For Use With (Application) | Food processors and contract labs, 5 mL coliform samples |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
S30722
|
3M™ Petrifilm™
6405 |
50 Pack | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
S30689
|
3M™ Petrifilm™
6415 |
500 Pack | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Coliform results in 24 hours
- Provide accurate, efficient, consistent results for food processors and contract labs
- Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
- Unique plate design to trap gas produce by coliform
- This plate has been awarded Official Methods of Analysis (OMA) #996.02 for Dairy products and NF VALIDATION certificate: 3M-01/7-03/99 for Human food products by AFNOR Certification
Specifications
Coliform | |
Count Plate | |
Environmental Test | |
Food processors and contract labs, 5 mL coliform samples |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title