Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Scientific High-Throughput Submarine Horizontal Electrophoresis Systems

Click to view available options
Gel Dimensions:
14 x 26 cm
40 x 26 cm
Description
CBS Economy Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.
Gel Tray Specifications:
- Winged gel trays for easy handling - no more sticking fingers into slippery buffer to extract gel tray
- Made from sturdy 6.4mm thick UV transparent acrylic
- Grooves in gel tray corners create agarose anchors to prevent gel flotation during electrophoresis
- Gel tray configuration allows single run or multiple simultaneous short runs
- Three options for gel casting; tapered baffles, external gel casters or continuously adjustable gel caster
- All combs are adjustable height and have an aluminum backing to keep the combs perfectly straight during gel casting with hot agarose
- All combs come with microtitre format to allow multi-channel pipetting for accuracy and speed
- Comb profiles are diagrammed at right to show many configurations, with or without markers
- PTFE-coated combs allow faster well formation and are recommended when using low percentage or low melt agarose
- Buffer recirculation ports allow maintenance of uniform pH and ionic strength and reduce frequency of buffer changes necessary during long runs.
- Tight-fitting safety cover reduces condensation during electrophoresis
- Coolant can be recirculated beneath gel bed to provide constant temperature runs
- Combs are printed with catalog number and thickness for easy identification
Applications:
Rapid Screening fo cloning products, rapid screening of restriction fragments, rapid screening of synthetic oligos, rapid screening of PCR products
Specifications
Specifications
Certifications/Compliance | CE |
Color | Blue |
Description | Compatible Gel Tray: WGT-3400 |
Type | Horizontal Gel Electrophoresis System |
Gel Dimensions | 40 x 26 cm |
Volume (Metric) Buffer | 3400 mL |
Warranty | 3 Years |
Dimensions (L x W) | Gel Tray I.D.: 40 x 24.7 cm (15.7 x 9.7 in.) |
Dimensions (L x W x H) | 64 x 28 x 10 cm (25.1 x 11 x 3.9 in.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction