Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HINFP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563921 Supplier Invitrogen™ Supplier No. PA563921
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHPRFRYKEH EDGYMRLQLV RYESVELTQQ LLRQPQEGSG LGTSLNESSL QGIILETVPG EPGRKE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

MIZF interacts with methyl-CpG-binding protein-2 (MBD2; MIM 603547), a component of the MeCP1 histone deacetylase (HDAC) complex, and plays a role in DNA methylation and transcription repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HINFP
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HINFP
Gene Accession No. Q9BQA5
Gene Alias AA589481; DKFZp434F162; Hinfp; HiNF-P; histone H4 gene-specific protein HiNF-P; histone H4 transcription factor; Histone nuclear factor P; MBD2 (methyl-CpG-binding protein)-interacting zinc finger protein; MBD2-interacting zinc finger; MBD2-interacting zinc finger 1; MBD2-interacting zinc finger protein; Methyl-CpG-binding protein 2-interacting zinc finger protein; MIZF; RGD1311492; ZNF743
Gene Symbols HINFP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HINFP. Recombinant protein control fragment (Product #RP-101943).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.