Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP1 Antibody (1B11), Novus Biologicals™
SDP

Catalog No. NB300204SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300204SS 0.025 mL
NB300204 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300204SS Supplier Novus Biologicals Supplier No. NB300204SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

HIP1 Monoclonal specifically detects HIP1 in Bacteria, Human, Mouse, Rat (Negative) samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen HIP1
Applications Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1B11
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23272104)
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. O00291
Gene Alias HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506
Gene Symbols HIP1
Host Species Mouse
Immunogen Human 3'-HIP1 (∼65 kDa)
Molecular Weight of Antigen 115 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3092
Test Specificity NB300-204 is specific for human HIP1. Does not cross-react with HIP1r.
Target Species Bacteria, Human, Mouse, Rat (Negative)
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.