Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP1 Antibody (1B11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 7 publications

$181.90 - $487.50

Specifications

Antigen HIP1
Clone 1B11
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23272104)
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300204SS
SDP
View Documents
Novus Biologicals
NB300204SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NB300204
SDP
View Documents
Novus Biologicals
NB300204
0.1 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

HIP1 Monoclonal specifically detects HIP1 in Human, Mouse, Bacteria, Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

HIP1
Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported in scientific literature (PMID 23272104)
Monoclonal
Ascites
RUO
Human, Mouse, Bacteria, Rat (Negative)
O00291
3092
Human 3'-HIP1 (∼65 kDa)
Primary
Store at -20C. Avoid freeze-thaw cycles.
115 kDa
1B11
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Neurodegeneration, Neuronal Cell Markers, Neuroscience
Ascites with 0.1% Sodium Azide
HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506
HIP1
IgG1 κ
Unpurified
NB300-204 is specific for human HIP1. Does not cross-react with HIP1r.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.