Learn More
Description
Specifications
Specifications
| Antigen | HIP1 |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4B10 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10000, Chromatin Immunoprecipitation reported in scientific literature, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature, Chromatin Immunoprecipitation (ChIP) |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | O00291 |
| Gene Alias | HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506 |
| Gene Symbols | HIP1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
