Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIP1 Antibody (4B10), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 9 publications
$206.50 - $499.50
Specifications
| Antigen | HIP1 |
|---|---|
| Clone | 4B10 |
| Dilution | Western Blot 1:10000, Chromatin Immunoprecipitation reported in scientific literature, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature, Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300203SS
![]() |
Novus Biologicals
NB300203SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB300203
![]() |
Novus Biologicals
NB300203 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
HIP1 Monoclonal specifically detects HIP1 in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| HIP1 | |
| Western Blot 1:10000, Chromatin Immunoprecipitation reported in scientific literature, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature, Chromatin Immunoprecipitation (ChIP) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat (Negative) | |
| O00291 | |
| 3092 | |
| Human 3'-HIP1 (∼65 kDa) | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 115 kDa |
| 4B10 | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| Ascites with 0.1% Sodium Azide | |
| HIP-1, HIP-I, huntingtin interacting protein 1, huntingtin-interacting protein 1, Huntingtin-interacting protein I, ILWEQ, MGC126506 | |
| HIP1 | |
| IgG1 κ | |
| Unpurified | |
| Human and Mouse. Does not react with rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title