Learn More
Description
Specifications
Specifications
| Antigen | HIP1 Related |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 10 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | O75146 |
| Gene Alias | FLJ14000, HIP-12, HIP12HIP1-related protein, HIP3, huntingtin interacting protein 1 related, huntingtin interacting protein 12, Huntingtin-interacting protein 12, huntingtin-interacting protein 1-related protein, ILWEQ, KIAA0655FLJ27022, MGC47513 |
| Gene Symbols | HIP1R |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
