Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP1 Related Antibody (1E1), Novus Biologicals™
SDP

Catalog No. NB300206SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300206SS 0.025 mL
NB300206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300206SS Supplier Novus Biologicals Supplier No. NB300206SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

HIP1 Related Monoclonal specifically detects HIP1 Related in Bacteria, Human, Mouse, Rat samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen HIP1 Related
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry reported in scientific literature, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. O75146
Gene Alias FLJ14000, HIP-12, HIP12HIP1-related protein, HIP3, huntingtin interacting protein 1 related, huntingtin interacting protein 12, Huntingtin-interacting protein 12, huntingtin-interacting protein 1-related protein, ILWEQ, KIAA0655FLJ27022, MGC47513
Gene Symbols HIP1R
Host Species Mouse
Immunogen Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt# O75146]
Molecular Weight of Antigen 120 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9026
Target Species Bacteria, Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.