Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK1 Antibody (OTI5F1), FITC, Novus Biologicals™
SDP

Catalog No. NBP272460F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272460F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272460F Supplier Novus Biologicals Supplier No. NBP272460F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIPK1 Monoclonal specifically detects HIPK1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen HIPK1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI5F1
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, homeodomain interacting protein kinase 1, homeodomain interacting protein kinase 1-like protein, homeodomain-interacting protein kinase 1, KIAA0630MGC26642, MGC33446, MGC33548, Myak, Nbak2, nuclear body associated kinase 2b
Gene Symbols HIPK1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 439-697 of human HIPK1 (NP_852003) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 204851.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.