Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224521SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224521SS 0.025 mg
NBP224521 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224521SS Supplier Novus Biologicals Supplier No. NBP224521SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIPK3 Polyclonal specifically detects HIPK3 in Canine, Human, Mouse, Primate, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen HIPK3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 5-10 ug/ml, Immunocytochemistry 2-10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9H422
Gene Alias Androgen receptor-interacting nuclear protein kinase, DYRK6Fas-interacting serine/threonine-protein kinase, EC 2.7.11.1, FIST, FIST3ANPK, homeodomain interacting protein kinase 3, homeodomain-interacting protein kinase 3, PKYHomolog of protein kinase YAK1, YAK1
Gene Symbols HIPK3
Host Species Rabbit
Immunogen A portion of amino acids 850-900 of human HIPK3 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10114
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_c) and chimpanzee, 94% homologous in orangutan and rhesus monkey, 83% homologous in horse and mouse, 77% homologous in cow and 47% homologous in rat.
Target Species Canine, Human, Mouse, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.