Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPK3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337419 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337419 Supplier Novus Biologicals Supplier No. NBP224521MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIPK3 Polyclonal antibody specifically detects HIPK3 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen HIPK3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Androgen receptor-interacting nuclear protein kinase, DYRK6Fas-interacting serine/threonine-protein kinase, EC 2.7.11.1, FIST, FIST3ANPK, homeodomain interacting protein kinase 3, homeodomain-interacting protein kinase 3, PKYHomolog of protein kinase YAK1, YAK1
Host Species Rabbit
Immunogen A portion of amino acids 850-900 of human HIPK3 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10114
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.