Learn More
Promega HIPK3 Kinase Enzyme System, 1mg
Catalog No. PRV6397 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6397 1 mg
1 options
Catalog No. PRV6397 Supplier Promega Supplier No. V6397
Only null left
Add to Cart
Add to Cart
Request a Quote
Recombinant human HIPK3 (163-562) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. HIPK3 (homeodomain interacting protein kinase 3) is a member of the HIPK family and is involved in apoptosis.
  • Easily Screen and Profile HIPK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C