Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIPPI Antibody (OTI4F6), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271985G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271985G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271985G Supplier Novus Biologicals Supplier No. NBP271985G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIPPI Monoclonal specifically detects HIPPI in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HIPPI
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4F6
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Dermal papilla-derived protein 8, DERP8, ESRRBL1huntingtin interacting protein-1 interacting protein, estrogen-related receptor beta like 1, Estrogen-related receptor beta-like protein 1, FLJ10147, HIPPIHIP1-interacting protein, intraflagellar transport 57 homolog (Chlamydomonas), intraflagellar transport protein 57 homolog, MHS4R2HIP1 protein interactor
Gene Symbols IFT57
Host Species Mouse
Immunogen Full length recombinant protein of human HIPPI (NP_060480) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55081.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.