Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hippocalcin Rabbit anti-Human, Mouse, Rat, Clone: 6N2A6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31586420UL
Description
Hippocalcin Monoclonal antibody specifically detects Hippocalcin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Hippocalcin | |
Monoclonal | |
Unconjugated | |
BDR2, Calcium-binding protein BDR-2, hippocalcin, neuron specific calcium-binding protein hippocalcin, neuron-specific calcium-binding protein hippocalcin | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Hippocalcin (P84074). | |
20 μg | |
Neuroscience | |
3208 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
6N2A6 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction