Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hippocalcin Rabbit anti-Human, Mouse, Rat, Clone: 6N2A6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Hippocalcin |
---|---|
Clone | 6N2A6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Hippocalcin Monoclonal antibody specifically detects Hippocalcin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Hippocalcin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3208 | |
IgG | |
Affinity purified |
6N2A6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BDR2, Calcium-binding protein BDR-2, hippocalcin, neuron specific calcium-binding protein hippocalcin, neuron-specific calcium-binding protein hippocalcin | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Hippocalcin (P84074). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title