Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIRA Rabbit anti-Human, Clone: 6Q7Y6, Novus Biologicals™
SDP

Catalog No. NB168524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168524 20 μg
NB168523 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168524 Supplier Novus Biologicals Supplier No. NBP31671620UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HIRA Monoclonal antibody specifically detects HIRA in Human samples. It is validated for Western Blot

Specifications

Antigen HIRA
Applications Western Blot
Classification Monoclonal
Clone 6Q7Y6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DGCR1DiGeorge critical region gene 1, HIR, HIR (histone cell cycle regulation defective) homolog A (S. cerevisiae), HIR histone cell cycle regulation defective homolog A (S. cerevisiae), TUP1, TUP1-like enhancer of split protein 1, TUPLE1protein HIRA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-675 of human HIRA (P54198).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Chromatin Modifiers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7290
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.