Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (3D5) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25263725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25263725 0.025 mg
NBP25263702 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25263725 Supplier Novus Biologicals Supplier No. NBP2526370.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

His Tag Monoclonal specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, Simple Western, ELISA.

Specifications

Antigen His Tag
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D5
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Host Species Rabbit
Immunogen Purified his tagged Fab fragment.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins.
Target Species Non-species specific
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.