Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (6HIS/6402R), HRP, Novus Biologicals™
SDP

Catalog No. NB381163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381163 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381163 Supplier Novus Biologicals Supplier No. NBP308360H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen His Tag
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 6HIS/6402R
Conjugate HRP
Formulation PBS
Gene Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Host Species Rabbit
Immunogen Hexa-histidine tagged human CD70 recombinant protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.