Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (AD1.1.10), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10064768U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064768U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064768U Supplier Novus Biologicals Supplier No. NB10064768UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

His Tag Monoclonal specifically detects His Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro Assay, Western Blot.

Specifications

Antigen His Tag
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro Assay, Western Blot
Classification Monoclonal
Clone AD1.1.10
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Host Species Mouse
Immunogen PAX6 transcription factor linked to histidine tag
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Test Specificity This antibody is reactive to histidine tags.
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.