Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (AD1.1.10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB297573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297573 Supplier Novus Biologicals Supplier No. FAB050MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Broad Species Reactivity samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Affinity Purification, CyTOF

Specifications

Antigen His Tag
Applications Western Blot, Flow Cytometry, Affinity Purification, CyTOF
Classification Monoclonal
Clone AD1.1.10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Flow Cytometry, Immunoaffinity Purification, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Host Species Mouse
Immunogen His-tagged peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.