Learn More
His Tag Antibody (AD1.1.10) - Non-Recombinant Monoclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | His Tag |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro Assay, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | AD1.1.10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:250, Flow Cytometry 1:10 - 1:50, ELISA 1:100 - 1:500, Immunohistochemistry 1:100 - 1:300, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:100 - 1:300, In vitro assay |
| Gene Alias | 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine |
| Host Species | Mouse |
| Immunogen | PAX6 transcription factor linked to histidine tag |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.