Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (AD1.1.10R), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB297571 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297571 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297571 Supplier Novus Biologicals Supplier No. FAB050RZ
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

His Tag Monoclonal specifically detects His Tag in Multi-species samples. It is validated for Flow Cytometry, Immunoaffinity Purification, Western Blot.

Specifications

Antigen His Tag
Applications Flow Cytometry, Immunoaffinity Chromatography, Western Blot
Classification Monoclonal
Clone AD1.1.10R
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunoaffinity Purification
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Host Species Mouse
Immunogen His-tagged peptide,
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.