Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB203956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203956 Supplier Novus Biologicals Supplier No. NBP125939G
Add to Cart
Add to Cart

Goat Polyclonal Antibody

His Tag Polyclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen His Tag
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Host Species Goat
Immunogen This His Tag Antibody was developed against Keyhole limpet haemocyanin (KLH) conjugated H-H-H-H-H-H.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.