Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (HIS.H8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB295277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295277 Supplier Novus Biologicals Supplier No. NBP231055AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF

Specifications

Antigen His Tag
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Classification Monoclonal
Clone HIS.H8
Conjugate Alexa Fluor 700
Dilution Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Host Species Mouse
Immunogen This His Tag Antibody (HIS.H8) was developed against a 6x His synthetic peptide.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.