Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (HIS.H8), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N23105PECY5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N23105PECY5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N23105PECY5 Supplier Novus Biologicals Supplier No. NBP231055PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Flow Cytometry.

Specifications

Antigen His Tag
Applications Flow Cytometry
Classification Monoclonal
Clone HIS.H8
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Host Species Mouse
Immunogen 6x His synthetic peptide.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.