Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (His17), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB029768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029768 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB029768 Supplier Novus Biologicals Supplier No. NB10072978AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

His Tag Monoclonal specifically detects His Tag in Tag samples. It is validated for ELISA, Immunoprecipitation, Western Blot.

Specifications

Antigen His Tag
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone His17
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with His6-Tag conjugated to BSA.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.