Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His Tag Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$606.50
Specifications
Antigen | His Tag |
---|---|
Clone | AD1.1.10R |
Dilution | Western Blot 0.2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoaffinity Purification |
Applications | Western Blot, Flow Cytometry, Affinity Purification |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB050R100
![]() |
R&D Systems
MAB050R100 |
100 μg |
Each of 1 for $606.50
|
|
|||||
Description
His Tag Monoclonal specifically detects His Tag in Multi-Species samples. It is validated for Western Blot, Flow Cytometry, Immunoaffinity Purification.Specifications
His Tag | |
Western Blot 0.2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoaffinity Purification | |
Monoclonal | |
Purified | |
RUO | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, polyHistidine, Poly-histidine | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Detects proteins containing accessible consecutive histidine regions. The antibody detects His tags located at the amino- or carboxyl- terminus. |
AD1.1.10R | |
Western Blot, Flow Cytometry, Affinity Purification | |
Unconjugated | |
Mouse | |
Multi-species | |
His-tagged peptide | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title