Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Antibody (RMH01), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB298812 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298812 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB298812 Supplier Novus Biologicals Supplier No. NBP326048V
Add to Cart
Add to Cart

Human Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, Western Blot, ELISA, ELISA, ELISA

Specifications

Antigen His Tag
Applications Western Blot, ELISA, ELISA, ELISA
Classification Monoclonal
Clone RMH01
Conjugate DyLight 405
Dilution Western Blot, Simple Western, ELISA, Sandwich ELISA Capture, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
Host Species Human
Immunogen This His Tag Antibody (RMH01) was developed against a mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis Tag at the C-terminus. This antibody is a Rabbit-Human Chimeric antibody, made by fusion of the antigen binding region (variable domains of the heavy and light chains, VH and VL) of NBP2-61482, with the constant domain of Human IgG1. The antibody is recognized only by an anti-Human IgG secondary antibody.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Epitope Tags
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.