Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Mouse anti-Nonspecies specific, DyLight 350, Clone: 33D10.D2, Novus Biologicals™
SDP

Catalog No. NB029796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029796 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB029796 Supplier Novus Biologicals Supplier No. NBP142781UV

Mouse Monoclonal Antibody

His Tag Monoclonal antibody specifically detects His Tag in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ELISA

Specifications

Antigen His Tag
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 33D10.D2
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Host Species Mouse
Immunogen His Tag antibody was produced in mice by repeated immunizations with 6X His epitope tag peptide H-H-H-H-H-H conjugated to KLH using maleimide.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.